Assay Details
| Gene Symbol: VPS35L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046368.2 | NP_001039833.2 | 3 - 4 | 298 | 67 | |
| GenBank mRNA | BC105446.1 | - | 3 - 4 | 334 | 67 | |
| EE353040.1 | - | 3 - 4 | 298 | 67 | ||
More Information
Set Membership: |
|

VPS35L
Genomic Map