Assay Details
| Gene Symbol: PGS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046395.2 | NP_001039860.2 | 9 - 10 | 1693 | 75 | |
| GenBank mRNA | BC105551.1 | - | 9 - 10 | 1692 | 75 | |
More Information
Set Membership: |
|

PGS1
Genomic Map