Assay Details
Gene Symbol: LDHC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046422.2 | NP_001039887.2 | 2 - 3 | 410 | 59 | |
NM_001113249.1 | NP_001106720.1 | 2 - 3 | 410 | 59 | ||
GenBank mRNA | BC105361.1 | - | 2 - 3 | 393 | 59 | |
DT830273.1 | - | 2 - 3 | 196 | 59 | ||
DT847039.1 | - | 2 - 3 | 414 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
