Assay Details
Gene Symbol: TARDBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046485.2 | NP_001039950.2 | 2 - 3 | 268 | 143 | |
GenBank mRNA | BC105527.1 | - | 2 - 3 | 324 | 143 | |
EE363761.1 | - | 2 - 3 | 268 | 143 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
