Assay Details
| Gene Symbol: ARL2BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076274.2 | NP_001069742.2 | 1 - 2 | 317 | 71 | |
| GenBank mRNA | BC134728.1 | - | 1 - 2 | 317 | 71 | |
More Information
Set Membership: |
|

ARL2BP
Genomic Map