Assay Details
| Gene Symbol: HPS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BF075133.1 | - | 4 - 5 | 351 | 84 | |
| BM087325.1 | - | 1 - 2 | 85 | 84 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HPS4| Gene Symbol: HPS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BF075133.1 | - | 4 - 5 | 351 | 84 | |
| BM087325.1 | - | 1 - 2 | 85 | 84 | ||
Set Membership: |
|