Assay Details
| Gene Symbol: CIB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076455.2 | NP_001069923.2 | 1 - 2 | 106 | 63 | |
| GenBank mRNA | BC109857.1 | - | 1 - 2 | 96 | 63 | |
| DT834350.1 | - | 1 - 2 | 107 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC109857.1 transcripts.
Set Membership: |
|

CIB4
Genomic Map