Assay Details
| Gene Symbol: TFB1M | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076896.2 | NP_001070364.1 | 4 - 5 | 597 | 70 | |
| NM_001114610.1 | NP_001108082.1 | 4 - 5 | 597 | 70 | ||
| GenBank mRNA | BC109841.1 | - | 4 - 5 | 585 | 70 | |
| DT811342.1 | - | 4 - 5 | 590 | 70 | ||
| DY454254.1 | - | 4 - 5 | 601 | 70 | ||
More Information
Set Membership: |
|

TFB1M
Genomic Map