Assay Details
Gene Symbol: SQLE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098061.1 | NP_001091530.1 | 1 - 2 | 376 | 92 | |
GenBank mRNA | BC142144.1 | - | 1 - 2 | 335 | 92 | |
BT030527.1 | - | 1 - 2 | 376 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
