Assay Details
Gene Symbol: POLDIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098103.1 | NP_001091572.1 | 7 - 8 | 779 | 69 | |
GenBank mRNA | BC146200.1 | - | 7 - 8 | 815 | 69 | |
BT030598.1 | - | 7 - 8 | 779 | 69 | ||
BT030703.1 | - | 7 - 8 | 752 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
