Assay Details
| Gene Symbol: LPAR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098105.1 | NP_001091574.1 | 2 - 3 | 246 | 78 | |
| GenBank mRNA | BC134782.1 | - | 2 - 3 | 246 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC134782.1, NM_001098105.1 transcripts.
Set Membership: |
|

LPAR4
Genomic Map