Assay Details
| Gene Symbol: PCDH12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098111.1 | NP_001091580.1 | 2 - 3 | 3009 | 89 | |
| GenBank mRNA | BC140692.1 | - | 2 - 3 | 3009 | 89 | |
More Information
Set Membership: |
|

PCDH12
Genomic Map