Assay Details
| Gene Symbol: RSAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098136.1 | NP_001091605.1 | 1 - 2 | 233 | 55 | |
| GenBank mRNA | BC140491.1 | - | 1 - 2 | 233 | 55 | |
More Information
Set Membership: |
|

RSAD1
Genomic Map