Assay Details
| Gene Symbol: ABCD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099037.1 | NP_001092507.1 | 1 - 2 | 1122 | 150 | |
| GenBank mRNA | BC146198.1 | - | 1 - 2 | 1122 | 150 | |
More Information
Set Membership: |
|

ABCD2
Genomic Map