Assay Details
| Gene Symbol: S100A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099042.2 | NP_001092512.1 | 1 - 2 | 280 | 86 | |
| GenBank mRNA | BC141991.1 | - | 1 - 2 | 281 | 86 | |
| BC148019.1 | - | 1 - 2 | 84 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC141991.1, BC148019.1, NM_001099042.2 transcripts.
Set Membership: |
|

S100A1
Genomic Map