Assay Details
| Gene Symbol: S100A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099042.2 | NP_001092512.1 | 2 - 3 | 437 | 119 | |
| GenBank mRNA | BC141991.1 | - | 2 - 3 | 438 | 119 | |
| BC148019.1 | - | 2 - 3 | 241 | 119 | ||
More Information
Set Membership: |
|

S100A1
Genomic Map