Assay Details
Gene Symbol: TNFRSF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099043.2 | NP_001092513.1 | 4 - 5 | 478 | 70 | |
GenBank mRNA | BC142115.1 | - | 4 - 5 | 479 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
