Assay Details
| Gene Symbol: PRSS54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099044.2 | NP_001092514.1 | 6 - 7 | 1060 | 66 | |
| GenBank mRNA | BC142059.1 | - | 6 - 7 | 1061 | 66 | |
More Information
Set Membership: |
|

PRSS54
Genomic Map