Assay Details
Gene Symbol: TOR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099058.2 | NP_001092528.1 | 2 - 3 | 502 | 62 | |
GenBank mRNA | BC142387.1 | - | 2 - 3 | 503 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
