Assay Details
| Gene Symbol: BRI3BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099087.1 | NP_001092557.1 | 2 - 3 | 417 | 60 | |
| GenBank mRNA | BC142091.1 | - | 2 - 3 | 417 | 60 | |
More Information
Set Membership: |
|

BRI3BP
Genomic Map