Assay Details
| Gene Symbol: GZMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099095.1 | NP_001092565.1 | 4 - 5 | 662 | 107 | |
| GenBank mRNA | BC142336.1 | - | 4 - 5 | 662 | 107 | |
More Information
Set Membership: |
|

GZMA
Genomic Map