Assay Details
Gene Symbol: PIMREG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099109.1 | NP_001092579.1 | 2 - 3 | 631 | 74 | |
GenBank mRNA | BC142479.1 | - | 2 - 3 | 631 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
