Assay Details
| Gene Symbol: UBE2W | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099122.3 | NP_001092592.1 | 3 - 4 | 251 | 123 | |
| GenBank mRNA | BC146162.1 | - | 3 - 4 | 220 | 123 | |
| FE011537.1 | - | 3 - 4 | 251 | 123 | ||
More Information
Set Membership: |
|

UBE2W
Genomic Map