Assay Details
| Gene Symbol: DDT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099150.1 | NP_001092620.1 | 1 - 2 | 152 | 78 | |
| GenBank mRNA | BC142376.1 | - | 1 - 2 | 152 | 78 | |
More Information
Set Membership: |
|

DDT
Genomic Map