Assay Details
Gene Symbol: RAB22A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099165.2 | NP_001092635.1 | 3 - 4 | 455 | 61 | |
GenBank mRNA | BC142190.1 | - | 3 - 4 | 456 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
