Assay Details
| Gene Symbol: PPP1R14D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099169.2 | NP_001092639.1 | 1 - 2 | 482 | 73 | |
| GenBank mRNA | BC142353.1 | - | 1 - 2 | 483 | 73 | |
More Information
Set Membership: |
|

PPP1R14D
Genomic Map