Assay Details
Gene Symbol: CDK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099178.1 | NP_001092648.1 | 4 - 5 | 345 | 90 | |
GenBank mRNA | BC142140.1 | - | 4 - 5 | 345 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
