Assay Details
Gene Symbol: CDK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099178.1 | NP_001092648.1 | 5 - 6 | 525 | 70 | |
GenBank mRNA | BC142140.1 | - | 5 - 6 | 525 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
