Assay Details
| Gene Symbol: RAC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099179.2 | NP_001092649.1 | 4 - 5 | 359 | 68 | |
| GenBank mRNA | BC142381.1 | - | 4 - 5 | 360 | 68 | |
More Information
Set Membership: |
|

RAC3
Genomic Map