Assay Details
| Gene Symbol: BCL7C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099722.2 | NP_001093192.1 | 3 - 4 | 390 | 138 | |
| GenBank mRNA | BC142100.1 | - | 3 - 4 | 391 | 138 | |
More Information
Set Membership: |
|

BCL7C
Genomic Map