Assay Details
| Gene Symbol: BCL7C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099722.2 | NP_001093192.1 | 4 - 5 | 560 | 107 | |
| GenBank mRNA | BC142100.1 | - | 4 - 5 | 561 | 107 | |
More Information
Set Membership: |
|

BCL7C
Genomic Map