Assay Details
Gene Symbol: TPM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101162.1 | NP_001094632.1 | 2 - 3 | 272 | 59 | |
GenBank mRNA | BC150074.1 | - | 2 - 3 | 272 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
