Assay Details
| Gene Symbol: EGLN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101164.2 | NP_001094634.1 | 1 - 2 | 630 | 67 | |
| GenBank mRNA | BC151442.1 | - | 1 - 2 | 631 | 67 | |
More Information
Set Membership: |
|

EGLN3
Genomic Map