Assay Details
Gene Symbol: EGLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101164.2 | NP_001094634.1 | 2 - 3 | 744 | 97 | |
GenBank mRNA | BC151442.1 | - | 2 - 3 | 745 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
