Assay Details
| Gene Symbol: CNOT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101167.2 | NP_001094637.1 | 1 - 2 | 147 | 109 | |
| GenBank mRNA | BC151334.1 | - | 1 - 2 | 148 | 109 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC151334.1, NM_001101167.2 transcripts.
Set Membership: |
|

CNOT9
Genomic Map