Assay Details
| Gene Symbol: SPACA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101193.2 | NP_001094663.1 | 1 - 2 | 106 | 92 | |
| GenBank mRNA | BC146122.1 | - | 1 - 2 | 107 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC146122.1, NM_001101193.2 transcripts.
Set Membership: |
|

SPACA9
Genomic Map