Assay Details
| Gene Symbol: AQP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101199.1 | NP_001094669.1 | 2 - 3 | 615 | 71 | |
| GenBank mRNA | BC151512.1 | - | 2 - 3 | 615 | 71 | |
More Information
Set Membership: |
|

AQP2
Genomic Map