Assay Details
| Gene Symbol: FAM181B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101223.2 | NP_001094693.1 | 1 | 1645 | 94 | |
| GenBank mRNA | BC151312.1 | - | 1 | 1646 | 94 | |
More Information
Set Membership: |
|

FAM181B
Genomic Map