Assay Details
| Gene Symbol: KCTD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101224.1 | NP_001094694.1 | 1 - 2 | 232 | 83 | |
| GenBank mRNA | BC148074.1 | - | 1 - 2 | 232 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC148074.1, NM_001101224.1 transcripts.
Set Membership: |
|

KCTD6
Genomic Map