Assay Details
| Gene Symbol: GCNT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101227.1 | NP_001094697.1 | 1 | 946 | 79 | |
| GenBank mRNA | BC146070.1 | - | 1 | 946 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GCNT2| Gene Symbol: GCNT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101227.1 | NP_001094697.1 | 1 | 946 | 79 | |
| GenBank mRNA | BC146070.1 | - | 1 | 946 | 79 | |
Set Membership: |
|