Assay Details
| Gene Symbol: BRINP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102140.2 | NP_001095610.1 | 7 - 8 | 1668 | 80 | |
| GenBank mRNA | BC151303.1 | - | 7 - 8 | 1669 | 80 | |
More Information
Set Membership: |
|

BRINP3
Genomic Map