Assay Details
| Gene Symbol: PSAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102150.1 | NP_001095620.1 | 6 - 7 | 822 | 67 | |
| GenBank mRNA | BC150088.1 | - | 6 - 7 | 822 | 67 | |
More Information
Set Membership: |
|

PSAT1
Genomic Map