Assay Details
| Gene Symbol: NEMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102161.1 | NP_001095631.1 | 5 - 6 | 651 | 92 | |
| GenBank mRNA | BC151486.1 | - | 5 - 6 | 651 | 92 | |
More Information
Set Membership: |
|

NEMP1
Genomic Map