Assay Details
| Gene Symbol: ANO10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102169.1 | NP_001095639.1 | 3 - 4 | 441 | 64 | |
| GenBank mRNA | BC151532.1 | - | 3 - 4 | 441 | 64 | |
More Information
Set Membership: |
|

ANO10
Genomic Map