Assay Details
| Gene Symbol: IL20RB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102175.2 | NP_001095645.1 | 1 - 2 | 250 | 79 | |
| GenBank mRNA | BC148928.1 | - | 1 - 2 | 251 | 79 | |
More Information
Set Membership: |
|

IL20RB
Genomic Map