Assay Details
| Gene Symbol: EGLN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102193.1 | NP_001095663.1 | 1 - 2 | 986 | 87 | |
| GenBank mRNA | BT030693.1 | - | 1 - 2 | 986 | 87 | |
More Information
Set Membership: |
|

EGLN2
Genomic Map