Assay Details
| Gene Symbol: ACTR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102213.1 | NP_001095683.1 | 6 - 7 | 800 | 90 | |
| GenBank mRNA | BC151356.1 | - | 6 - 7 | 800 | 90 | |
More Information
Set Membership: |
|

ACTR2
Genomic Map