Assay Details
Gene Symbol: ZNF565 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102216.2 | NP_001095686.1 | 1 - 2 | 41 | 91 | |
GenBank mRNA | BC149023.1 | - | 1 - 2 | 42 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC149023.1, NM_001102216.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
