Assay Details
Gene Symbol: CDYL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102223.2 | NP_001095693.1 | 2 - 3 | 943 | 54 | |
GenBank mRNA | BC151603.1 | - | 2 - 3 | 945 | 54 | |
EF687906.1 | - | 4 - 5 | 1026 | 54 | ||
EF690280.1 | - | 2 - 3 | 996 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
