Assay Details
| Gene Symbol: TMEM132A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102237.2 | NP_001095707.1 | 5 - 6 | 1155 | 61 | |
| GenBank mRNA | BC151451.1 | - | 5 - 6 | 1156 | 61 | |
| BC153245.1 | - | 5 - 6 | 1120 | 61 | ||
More Information
Set Membership: |
|

TMEM132A
Genomic Map